Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523989_m1
          See other CTU1 GE Assays ›
          Gene Symbol:
          Ctu1
          Gene Name:
          cytosolic thiouridylase subunit 1 homolog (S. pombe)
          Gene Aliases:
          Atpbd3, BC005752
          Chromosome Location:
          Chr.7: 43672031 - 43678298 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523989_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctu1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145582.1 NP_663557.1 2 - 3 593 64
          GenBank mRNA AK155284.1 - 2 - 3 608 64
          BC005752.1 - 2 - 3 593 2654430 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000768.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctu1

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble uridine modification
          tRNA processing
          biological_process
          tRNA thio-modification
          tRNA binding
          molecular_function
          telomerase activity
          RNA binding
          RNA helicase activity
          DNA primase activity
          sulfate adenylyltransferase activity
          RNA guanylyltransferase activity
          tRNA guanylyltransferase activity
          mannose-phosphate guanylyltransferase activity
          transferase activity
          DNA/RNA helicase activity
          CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
          phospholactate guanylyltransferase activity
          ATP:coenzyme F420 adenylyltransferase activity
          UDP-N-acetylgalactosamine diphosphorylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083585 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083571 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083584 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083580 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 BC005752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 AK155284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 AK160967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 NM_145582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 BC005752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 AK155284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 AK160967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114353 NM_145582.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475417 AK032241.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154197 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 BC005752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 AK155284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 AK160967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 NM_145582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 BC005752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 AK155284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 AK160967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114353 NM_145582.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154197 - -
          Whole Mouse Genome G4122A A_51_P114353 BC005752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 AK155284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 AK160967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 NM_145582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 BC005752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 AK155284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 AK160967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114353 NM_145582.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114511 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05920756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05920755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05920752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06399274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083585 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083571 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083584 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25083583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05937957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06451052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05939213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06555680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06098433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05939207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06555681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06127985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06381523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05976145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06470627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552460 - -
          Affymetrix HT MG-430 PM Array Plate 1424023_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552458 - -
          Mouse Genome 430 2.0 Array 1424023_at - -
          Mouse Genome 430 2.0 Array 1424023_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233189_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233189_at - -
          Affymetrix Mouse Clariom S Array PSR0700006239.mm.1 TC0700000768.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700006239.mm.1 NM_145582.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700006239.mm.1 NM_145582.1 Refseq
          Mouse Expression Set 430 1424023_PM_at - -
          Mouse Expression Set 430 1424023_at - -
          Murine Genome U74v2 Set 131153_PM_at - -
          Murine Genome U74v2 Set 131153_at - -
          Mouse Genome 430A 2.0 Array 1424023_PM_at - -
          Mouse Genome 430A 2.0 Array 1424023_at - -
          Affymetrix HT Mouse Genome 430A Array 1424023_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424023_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438449 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090739 - -
          Illumina mouse-6 v1.0 expression beadchip 1090739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249674 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249674 - -
          mouse-6 v1.1 (Illumina) scl32747.3_60-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32747.3_60-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2783557 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2783562 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2783562 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2783557 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32747.3_60-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline