Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524836_m1
          See other SLC26A8 GE Assays ›
          Gene Symbol:
          Slc26a8
          Gene Name:
          solute carrier family 26, member 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 28637779 - 28691237 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524836_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc26a8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290320.1 NP_001277249.1 2 - 3 467 72
          GenBank mRNA BC027076.1 - 2 - 3 467 5361175 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001767.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          anion transport
          chloride transport
          multicellular organism development
          spermatogenesis
          sulfate transport
          oxalate transport
          cell differentiation
          sperm motility
          sperm capacitation
          meiotic cell cycle
          transmembrane transport
          sulfate transmembrane transport
          chloride transmembrane transport
          organic acid transmembrane transport
          chloride channel activity
          secondary active sulfate transmembrane transporter activity
          anion transmembrane transporter activity
          sulfate transmembrane transporter activity
          anion:anion antiporter activity
          oxalate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P22640 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P193189 BC027076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P193189 BC027076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193189 BC027076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193189 BC027076.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449497 - -
          Affymetrix HT MG-430 PM Array Plate 1425915_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439281_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449487 - -
          Mouse Genome 430 2.0 Array 1425915_at - -
          Mouse Genome 430 2.0 Array 1439281_PM_at - -
          Mouse Genome 430 2.0 Array 1425915_PM_at - -
          Mouse Genome 430 2.0 Array 1439281_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224661_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224661_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700016315.mm.1 TC1700001767.mm.1 MTA TC
          Mouse Expression Set 430 1425915_PM_at - -
          Mouse Expression Set 430 1425915_at - -
          Mouse Expression Set 430 1439281_PM_at - -
          Mouse Expression Set 430 1439281_at - -
          Mouse Genome 430A 2.0 Array 1425915_at - -
          Mouse Genome 430A 2.0 Array 1425915_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425915_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425915_at - -
          Affymetrix HT Mouse Genome 430B Array 1439281_at - -
          Affymetrix HT Mouse Genome 430B Array 1439281_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774932 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060403 - -
          MouseWG-6 v2 Expression BeadChip 6590725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604723 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604723 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604723 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230768 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline