Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525343_m1
          See other TCAF2 GE Assays ›
          Gene Symbol:
          Tcaf2
          Gene Name:
          TRPM8 channel-associated factor 2
          Gene Aliases:
          Fam115c
          Chromosome Location:
          Chr.6: 42623043 - 42645041 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525343_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcaf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146174.1 NP_666286.1 7 - 8 2680 76
          GenBank mRNA AK131169.1 - 7 - 8 2893 76
          BC008555.1 - 7 - 8 2697 3592166 76
          BC011487.1 - 7 - 8 2680 3499593 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002177.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcaf2

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          transport
          negative regulation of anion channel activity
          positive regulation of cell migration
          positive regulation of protein targeting to membrane
          ion channel binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 NM_146174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 AK131169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 BC008555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 BC011487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 NM_146174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 AK131169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 BC008555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432794 BC011487.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16988 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168695 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154106 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 NM_146174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 AK131169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 BC008555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 BC011487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 NM_146174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 AK131169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 BC008555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432794 BC011487.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399420 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01314368 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05878451 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01314340 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154106 - -
          Whole Mouse Genome G4122A A_51_P432794 NM_146174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 AK131169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 BC008555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 BC011487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 NM_146174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 AK131169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 BC008555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432794 BC011487.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129158 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154106 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05315094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05315090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05315093 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05315099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05315092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05682432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05690346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05315090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05269868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05614445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05315105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05690339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05315103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05654637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05315109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05315102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05690343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544453 - -
          Affymetrix HT MG-430 PM Array Plate 1424761_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10544452 - -
          Mouse Genome 430 2.0 Array 1424761_at - -
          Mouse Genome 430 2.0 Array 1424761_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232748_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232748_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600017154.mm.1 TC0600002177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017154.mm.1 NM_146174.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017154.mm.1 NM_146174.1 Refseq
          Mouse Expression Set 430 1424761_at - -
          Mouse Expression Set 430 1424761_PM_at - -
          Murine Genome U74v2 Set 130121_PM_at - -
          Murine Genome U74v2 Set 130121_at - -
          Mouse Genome 430A 2.0 Array 1424761_at - -
          Mouse Genome 430A 2.0 Array 1424761_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424761_at - -
          Affymetrix HT Mouse Genome 430A Array 1424761_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 709269 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150685 - -
          Illumina mouse-6 v1.0 expression beadchip 4150685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618272 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618272 - -
          mouse-6 v1.1 (Illumina) scl29071.7_155-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29071.7_155-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2985836 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2985836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline