Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525679_s1
          See other OLFR729 GE Assays ›
          Gene Symbol:
          Olfr729
          Gene Name:
          olfactory receptor 729
          Gene Aliases:
          MOR246-6
          Chromosome Location:
          Chr.14: 50147901 - 50148872 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525679_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr729
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146278.1 NP_666390.1 1 846 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001976.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr729

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027037 NM_146278.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101275 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027037 NM_146278.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P436092 NM_146278.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29596569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101275 - -
          Whole Mouse Genome G4122A A_51_P436092 NM_146278.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126685 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101275 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11636360 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11636357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11358429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11358436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11358432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11636359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29596577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29596598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419443 - -
          Affymetrix Mouse Gene 1.1 ST Array 10419442 - -
          Affymetrix Mouse Clariom S Array PSR1400014829.mm.1 NM_146278.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014829.mm.1 NM_146278.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014829.mm.1 TC1400001976.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 356876 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253398 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253398 - -
          mouse-6 v1.1 (Illumina) scl2040.1.1_302-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2040.1.1_302-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1253398 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253398 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline