Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526156_s1
          See other OLFR893 GE Assays ›
          Gene Symbol:
          Olfr893
          Gene Name:
          olfactory receptor 893
          Gene Aliases:
          MOR170-11
          Chromosome Location:
          Chr.9: 38209052 - 38209993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526156_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr893
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146336.2 NP_666448.2 1 471 72
          GenBank mRNA BC130242.1 - 1 472 40142233 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000390.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr893

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967578 NM_146336.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967578 BC130242.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187371 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164950 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967578 NM_146336.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967578 BC130242.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174505 NM_146336.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174505 BC130242.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26433993 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164950 - -
          Whole Mouse Genome G4122A A_51_P174505 NM_146336.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174505 BC130242.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115104 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26433993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26433978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584383 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584382 - -
          Affymetrix Mouse Clariom S Array PSR0900002782.mm.1 NM_146336.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002782.mm.1 NM_146336.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002782.mm.1 TC0900000390.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 606690 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237379 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237379 - -
          mouse-6 v1.1 (Illumina) scl7577.1.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7577.1.1_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237379 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline