Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526181_s1
          See other OLFR77 GE Assays ›
          Gene Symbol:
          Olfr77
          Gene Name:
          olfactory receptor 77
          Gene Aliases:
          18A, MOR143-1
          Chromosome Location:
          Chr.9: 19917229 - 19922510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526181_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146339.3 NP_666451.1 3 525 93
          GenBank mRNA BC046590.1 - 3 525 6309128 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr77

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183944 BC046590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183944 NM_146339.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112321 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124253 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165339 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150233 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183944 BC046590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183944 NM_146339.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180603 Y15526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P154922 BC046590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P154922 NM_146339.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P188899 BC046590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P188899 NM_146339.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165339 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124253 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06715516 - -
          Whole Mouse Genome G4122A A_51_P180603 Y15526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188899 BC046590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188899 NM_146339.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154922 BC046590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154922 NM_146339.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102535 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165339 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124253 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08007244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08007254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08007256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08007239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07963826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07669812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583430 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583428 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 357083 - -
          ABI Mouse Genome Survey Microarray 751839 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840008 - -
          Illumina mouse-6 v1.0 expression beadchip 840008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604160 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604160 - -
          mouse-6 v1.1 (Illumina) scl0258336.3_176-S - -
          Illumina Sentrix mouse-6.v1 bead array 840008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258336.3_176-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604160 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline