Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526714_s1
          See other OLFR868 GE Assays ›
          Gene Symbol:
          Olfr868
          Gene Name:
          olfactory receptor 868
          Gene Aliases:
          MOR145-4
          Chromosome Location:
          Chr.9: 20098626 - 20101690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526714_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr868
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146559.2 NP_666770.1 2 459 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000160.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr868

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144395 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341719 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144395 - -
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120823 NM_146559.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101564 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006065 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08006064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08076215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08076214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08162398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08006051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07964261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07678104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341716 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583435 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583434 - -
          Affymetrix Mouse Clariom S Array PSR0900001014.mm.1 NM_146559.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001014.mm.1 NM_146559.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001014.mm.1 TC0900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001015.mm.1 NM_146559.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001015.mm.1 NM_146559.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001015.mm.1 TC0900000160.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 811239 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3060500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734325 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734325 - -
          mouse-6 v1.1 (Illumina) scl7633.1.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7633.1.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2734325 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2734325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0