Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528563_s1
          See other OLFR908 GE Assays ›
          Gene Symbol:
          Olfr908
          Gene Name:
          olfactory receptor 908
          Gene Aliases:
          MOR165-2
          Chromosome Location:
          Chr.9: 38516034 - 38516965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528563_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr908
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146872.1 NP_667083.1 1 896 90
          GenBank mRNA BC112370.1 - 1 893 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr908

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225723 BC112370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225723 BC112370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175190 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225723 BC112370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225723 NM_146872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225723 BC112370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225723 BC112370.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225723 BC112370.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175190 - -
          Whole Mouse Genome G4122A A_51_P225723 BC112370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225723 NM_146872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225723 BC112370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225723 NM_146872.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125016 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07798604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07798599 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07798587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07798593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07798601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08110953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584423 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584422 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 330511 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6980670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2762445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2762445 - -
          mouse-6 v1.1 (Illumina) scl37091.1.141_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37091.1.141_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980670 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2813940 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2762444 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813940 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0