Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529069_s1
          See other OLFR353 GE Assays ›
          Gene Symbol:
          Olfr353
          Gene Name:
          olfactory receptor 353
          Gene Aliases:
          MOR127-3
          Chromosome Location:
          Chr.2: 36889911 - 36890846 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529069_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr353
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146941.1 NP_667152.1 1 664 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003387.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr353

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123029 NM_146941.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164660 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123029 NM_146941.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P280938 NM_146941.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164660 - -
          Whole Mouse Genome G4122A A_52_P280938 NM_146941.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126356 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01215654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01467080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02141380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01467081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01355972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01215650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01215652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01355958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01215660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10482126 - -
          Affymetrix Mouse Gene 1.1 ST Array 10482125 - -
          Affymetrix Mouse Clariom S Array PSR0200029108.mm.1 NM_146941.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029108.mm.1 NM_146941.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029108.mm.1 TC0200003387.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 841178 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6040091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705263 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705263 - -
          mouse-6 v1.1 (Illumina) scl4524.1.1_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4524.1.1_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2927593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2927593 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl4524.1.1_43-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline