Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530019_s1
          See other OLFR449 GE Assays ›
          Gene Symbol:
          Olfr449
          Gene Name:
          olfactory receptor 449
          Gene Aliases:
          MOR103-1
          Chromosome Location:
          Chr.6: 42834384 - 42838882 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530019_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr449
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147064.1 NP_667275.1 1 457 72
          GenBank mRNA BC138166.1 - 1 488 8861185 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000470.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr449

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180675 NM_147064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180675 BC138166.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156950 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136397 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149399 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180675 NM_147064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180675 BC138166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P359525 NM_147064.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P359525 BC138166.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400358 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149399 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156950 - -
          Whole Mouse Genome G4122A A_51_P359525 NM_147064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359525 BC138166.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123546 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149399 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156950 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136397 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05090997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05132134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05334823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05132138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05091001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05334825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05624829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05514752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05233991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05132134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05624812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400390 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537802 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537801 - -
          Affymetrix Mouse Clariom S Array PSR0600003739.mm.1 TC0600000470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003739.mm.1 NM_147064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003739.mm.1 NM_147064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003738.mm.1 TC0600000470.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 733358 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610463 - -
          Illumina mouse-6 v1.0 expression beadchip 610463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658882 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658882 - -
          mouse-6 v1.1 (Illumina) scl0259067.1_292-S - -
          Illumina Sentrix mouse-6.v1 bead array 610463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259067.1_292-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658882 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658882 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline