Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530122_s1
          See other OLFR615 GE Assays ›
          Gene Symbol:
          Olfr615
          Gene Name:
          olfactory receptor 615
          Gene Aliases:
          MOR19-2
          Chromosome Location:
          Chr.7: 103560479 - 103561420 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530122_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr615
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147080.2 NP_667291.1 1 451 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001498.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr615

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979412 NM_147080.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731141 AF069992.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731141 AK013207.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731141 AK029295.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731141 BC012960.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731141 NM_011276.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 180311 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102700 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979412 NM_147080.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017242 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279714 NM_147080.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102700 - -
          Whole Mouse Genome G4122A A_51_P279714 NM_147080.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128295 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06181832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06181815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555800 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555799 - -
          Affymetrix Mouse Clariom S Array PSR0700012760.mm.1 TC0700001498.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012760.mm.1 NM_147080.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012760.mm.1 NM_147080.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 587311 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 130451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231317 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231317 - -
          mouse-6 v1.1 (Illumina) scl32266.1.1_244-S - -
          Illumina Sentrix mouse-6.v1 bead array 130451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32266.1.1_244-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231317 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline