Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530308_s1
          See other OLFR552 GE Assays ›
          Gene Symbol:
          Olfr552
          Gene Name:
          olfactory receptor 552
          Gene Aliases:
          MOR28-1
          Chromosome Location:
          Chr.7: 102604356 - 102605309 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530308_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr552
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147102.2 NP_667313.2 1 251 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001461.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr552

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046278 NM_147102.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986361 NM_147102.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175384 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986361 NM_147102.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046278 NM_147102.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P414815 NM_147102.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175384 - -
          Whole Mouse Genome G4122A A_51_P414815 NM_147102.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104240 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116591 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06586185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06169591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06083157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06169592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06353425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05943746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379689 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555726 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555725 - -
          Affymetrix Mouse Clariom S Array PSR0700012686.mm.1 TC0700001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012686.mm.1 NM_147102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012686.mm.1 NM_147102.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 304550 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594364 - -
          mouse-6 v1.1 (Illumina) scl9414.1.1_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9414.1.1_197-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594364 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0