Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00548865_m1
          See other CCDC137 GE Assays ›
          Gene Symbol:
          Ccdc137
          Gene Name:
          coiled-coil domain containing 137
          Gene Aliases:
          3110023B02Rik
          Chromosome Location:
          Chr.11: 120458129 - 120464358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00548865_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc137
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152807.3 NP_690020.1 1 - 2 148 68
          GenBank mRNA AK156682.1 - 1 - 2 157 68
          BC026662.1 - 1 - 2 150 4989084 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002027.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc137

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184671 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998751 AK156682.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998751 BC026662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998751 AK013412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998751 NM_152807.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66352 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147976 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998751 AK156682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998751 BC026662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998751 AK013412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998751 NM_152807.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184687 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147976 - -
          Whole Mouse Genome G4122A A_51_P137383 BC026662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137383 AK013412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137383 NM_152807.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129416 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135247 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147976 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09480676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09480670 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09480680 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09616266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09616246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09616258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184683 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10383363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383364 - -
          Affymetrix HT MG-430 PM Array Plate 1424533_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10383360 - -
          Mouse Genome 430 2.0 Array 1424533_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424533_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67291_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67291_at - -
          Affymetrix Mouse Clariom S Array PSR1100019469.mm.1 NM_152807.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019469.mm.1 NM_152807.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019469.mm.1 TC1100002027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019467.mm.1 NM_152807.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019467.mm.1 NM_152807.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019467.mm.1 TC1100002027.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019468.mm.1 NM_152807.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019468.mm.1 NM_152807.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019468.mm.1 TC1100002027.mm.1 MTA TC
          Mouse Expression Set 430 1424533_a_at - -
          Mouse Expression Set 430 1424533_PM_a_at - -
          Murine Genome U74v2 Set 171607_PM_r_at - -
          Murine Genome U74v2 Set 171607_r_at - -
          Mouse Genome 430A 2.0 Array 1424533_a_at - -
          Mouse Genome 430A 2.0 Array 1424533_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424533_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424533_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 453514 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450500 - -
          Illumina mouse-6 v1.0 expression beadchip 5570692 - -
          Illumina mouse-6 v1.0 expression beadchip 2940619 - -
          Illumina mouse-6 v1.0 expression beadchip 1450500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603860 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217150 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603860 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217150 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220014 - -
          mouse-6 v1.1 (Illumina) scl067291.6_198-S - -
          mouse-6 v1.1 (Illumina) scl0067291.1_12-S - -
          mouse-6 v1.1 (Illumina) scl067291.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940619 - -
          Illumina Sentrix mouse-6.v1 bead array 1450500 - -
          Illumina Sentrix mouse-6.v1 bead array 5570692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067291.6_198-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067291.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067291.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940619 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570692 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217150 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220014 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2836246 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220014 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217150 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2836246 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067291.6_198-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline