Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549096_m1
          See other PPAT GE Assays ›
          Gene Symbol:
          Ppat
          Gene Name:
          phosphoribosyl pyrophosphate amidotransferase
          Gene Aliases:
          5730454C12Rik, AA408689, AA675351, AI507113, C79945
          Chromosome Location:
          Chr.5: 76913249 - 76951578 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549096_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172146.2 NP_742158.1 8 - 9 1508 64
          GenBank mRNA AK147750.1 - 8 - 9 1158 64
          AK148034.1 - 8 - 9 1112 64
          AK159141.1 - 9 - 10 1658 64
          AK159859.1 - 9 - 10 1658 64
          AK160542.1 - 8 - 9 1133 64
          AK164506.1 - 8 - 9 1122 64
          AK167057.1 - 8 - 9 1147 64
          BC006053.1 - 7 - 8 1350 64
          BC023841.1 - 8 - 9 1135 5346105 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002642.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppat

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glutamine catabolic process
          purine ribonucleoside monophosphate biosynthetic process
          ribose phosphate metabolic process
          protein homotetramerization
          amidophosphoribosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK148034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK159141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK159859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 NM_172146.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK017664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK148034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK159141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK159859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 NM_172146.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P296905 AK017664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 BC023841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 BC006053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK164506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK167057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK148034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 BC023841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 BC006053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK164506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK167057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659258 AK148034.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359614 NM_134236.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88339 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153336 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112685 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK148034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK159141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK159859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 NM_172146.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK017664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK148034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK159141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK159859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 NM_172146.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P296905 AK017664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 BC023841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 BC006053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK164506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK167057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK148034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 BC023841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 BC006053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK164506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK167057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659258 AK148034.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05624083 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112685 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05624095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01139190 - -
          Whole Mouse Genome G4122A A_52_P659258 BC023841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 BC006053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK164506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK167057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK148034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 BC023841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 BC006053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK164506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK167057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659258 AK148034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK148034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK159141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK159859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 NM_172146.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK017664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK148034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK159141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK159859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 NM_172146.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296905 AK017664.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107828 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112685 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23818920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23818924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23818928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04597424 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10530809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530813 - -
          Affymetrix HT MG-430 PM Array Plate 1428543_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441770_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452831_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10530806 - -
          Mouse Genome 430 2.0 Array 1441770_at - -
          Mouse Genome 430 2.0 Array 1452831_s_at - -
          Mouse Genome 430 2.0 Array 1452831_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428543_PM_at - -
          Mouse Genome 430 2.0 Array 1428543_at - -
          Mouse Genome 430 2.0 Array 1441770_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231327_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231327_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500023362.mm.1 TC0500002642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023362.mm.1 NM_172146.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023362.mm.1 NM_172146.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023361.mm.1 TC0500002642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023361.mm.1 NM_172146.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023361.mm.1 NM_172146.2 Refseq
          Mouse Expression Set 430 1452831_s_at - -
          Mouse Expression Set 430 1428543_PM_at - -
          Mouse Expression Set 430 1428543_at - -
          Mouse Expression Set 430 1452831_PM_s_at - -
          Mouse Expression Set 430 1441770_PM_at - -
          Mouse Expression Set 430 1441770_at - -
          Murine Genome U74v2 Set 95554_PM_at - -
          Murine Genome U74v2 Set 95554_at - -
          Murine Genome U74v2 Set 95553_PM_at - -
          Murine Genome U74v2 Set 95553_at - -
          Murine Genome U74v2 Set 168502_PM_r_at - -
          Murine Genome U74v2 Set 168502_r_at - -
          Mouse Genome 430A 2.0 Array 1452831_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452831_s_at - -
          Mouse Genome 430A 2.0 Array 1428543_at - -
          Mouse Genome 430A 2.0 Array 1428543_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428543_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452831_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452831_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1428543_at - -
          Affymetrix HT Mouse Genome 430B Array 1441770_at - -
          Affymetrix HT Mouse Genome 430B Array 1441770_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 469562 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980019 - -
          Illumina mouse-6 v1.0 expression beadchip 101410519 - -
          Illumina mouse-6 v1.0 expression beadchip 1980019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245424 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222130 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245424 - -
          mouse-6 v1.1 (Illumina) scl00231327.1_147-S - -
          mouse-6 v1.1 (Illumina) scl26453.11_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 101410519 - -
          Illumina Sentrix mouse-6.v1 bead array 1980019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00231327.1_147-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980019 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101410519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245424 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222130 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline