Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549843_m1
          See other DTX4 GE Assays ›
          Gene Symbol:
          Dtx4
          Gene Name:
          deltex 4 homolog (Drosophila)
          Gene Aliases:
          AI449438, BC044798, D930042H13, RNF155
          Chromosome Location:
          Chr.19: 12466329 - 12502201 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dtx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172442.3 NP_766030.3 7 - 8 2304 77
          GenBank mRNA AK086624.1 - 6 - 7 873 77
          BC044798.1 - 2 - 3 77
          BC058647.1 - 7 - 8 1762 6405881 77
          BC069975.1 - 6 - 7 844 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001163.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dtx4

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          Notch signaling pathway
          biological_process
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900010790.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010790.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010790.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010792.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010792.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010792.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010793.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010793.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010793.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010791.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010791.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010791.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010797.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010797.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010797.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010789.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010789.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010789.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010794.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010794.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010794.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010795.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010795.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010795.mm.1 TC1900001163.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010796.mm.1 NM_172442.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010796.mm.1 NM_172442.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010796.mm.1 TC1900001163.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0