Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00554956_m1
          See other CLEC9A GE Assays ›
          Gene Symbol:
          Clec9a
          Gene Name:
          C-type lectin domain family 9, member a
          Gene Aliases:
          9830005G06Rik, DNGR-1
          Chromosome Location:
          Chr.6: 129408862 - 129501701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00554956_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec9a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205363.1 NP_001192292.1 1 - 2 185 61
          NM_001205365.1 NP_001192294.1 1 - 2 185 61
          NM_172732.3 NP_766320.2 1 - 2 185 61
          GenBank mRNA AK036399.1 - 1 - 2 186 61
          AK041288.1 - 1 - 2 185 61
          BC087955.1 - 1 - 2 184 30119997 61
          EU339277.1 - 1 - 2 92 61
          EU339278.1 - 1 - 2 92 61
          EU339280.1 - 1 - 2 92 61
          EU339281.1 - 1 - 2 92 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003537.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec9a

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          positive regulation of cytokine secretion
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103027 AK036399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103027 AK041288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103027 BC087955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103027 BY542702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103027 NM_172732.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16074 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168973 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103027 AK036399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103027 AK041288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103027 BC087955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103027 BY542702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103027 NM_172732.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24843309 - -
          Whole Mouse Genome G4122A A_51_P308394 AK036399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124197 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103027 AK036399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103027 AK041288.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103027 BC087955.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103027 BY542702.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103027 NM_172732.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639631 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05468794 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05639623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05468788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05468784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05724122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05468778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05468790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05057951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05583871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843348 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542187 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542181 - -
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 TC0600003537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205363.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205363.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205365.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_001205365.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_172732.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012499.mm.1 NM_172732.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 TC0600003537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_001205363.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_001205363.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_001205365.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_001205365.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_172732.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012501.mm.1 NM_172732.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 559867 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100840014 - -
          MouseWG-6 v2 Expression BeadChip 940500 - -
          Illumina mouse-6 v1.0 expression beadchip 100840014 - -
          Illumina mouse-6 v1.0 expression beadchip 940500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237681 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239591 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237681 - -
          mouse-6 v1.1 (Illumina) scl29464.7_446-S - -
          Illumina Sentrix mouse-6.v1 bead array 940500 - -
          Illumina Sentrix mouse-6.v1 bead array 100840014 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29464.7_446-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100840014 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2929768 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237681 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239591 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2929768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline