Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00556590_s1
          See other 4933405O20RIK GE Assays ›
          Gene Symbol:
          4933405O20Rik
          Gene Name:
          RIKEN cDNA 4933405O20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 50599190 - 50600528 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00556590_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933405O20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172901.2 NP_766489.1 1 344 104
          GenBank mRNA AK077105.1 - 1 345 104
          BC060958.1 - 1 321 6744040 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000910.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4933405O20Rik

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          oxidation-reduction process
          nucleotide binding
          magnesium ion binding
          isocitrate dehydrogenase (NAD+) activity
          ATP binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          metal ion binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 AK077105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 BC060958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 NM_172901.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 AK077105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 BC060958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299964 NM_172901.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103207 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 AK077105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 BC060958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 NM_172901.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 AK077105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 BC060958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299964 NM_172901.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25119083 - -
          Whole Mouse Genome G4122A A_51_P299964 AK077105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299964 BC060958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299964 NM_172901.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299964 AK077105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299964 BC060958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299964 NM_172901.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06485212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06485213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06485225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25119081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06517117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06485216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06485212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06639720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06476896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25119064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10553472 - -
          Affymetrix HT MG-430 PM Array Plate 1437427_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10553471 - -
          Mouse Genome 430 2.0 Array 1437427_PM_at - -
          Mouse Genome 430 2.0 Array 1437427_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243996_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243996_at - -
          Affymetrix Mouse Clariom S Array PSR0700008228.mm.1 TC0700000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008228.mm.1 NM_172901.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008228.mm.1 NM_172901.2 Refseq
          Mouse Expression Set 430 1437427_at - -
          Mouse Expression Set 430 1437427_PM_at - -
          Murine Genome U74v2 Set 165844_PM_r_at - -
          Murine Genome U74v2 Set 169830_r_at - -
          Murine Genome U74v2 Set 165844_r_at - -
          Murine Genome U74v2 Set 169830_PM_r_at - -
          Murine Genome U74v2 Set 167695_PM_f_at - -
          Murine Genome U74v2 Set 167695_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1437427_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437427_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 756557 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2370670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229724 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229724 - -
          mouse-6 v1.1 (Illumina) scl32627.1.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32627.1.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370670 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229724 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229724 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0