Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00557832_m1
          See other ERICH5 GE Assays ›
          Gene Symbol:
          Erich5
          Gene Name:
          glutamate rich 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 34453312 - 34473892 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00557832_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erich5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173421.2 NP_775597.1 1 - 2 397 111
          GenBank mRNA AK135792.1 - 1 - 2 128 111
          AK162262.1 - 1 - 2 155 111
          BC030476.1 - 1 - 2 405 5368170 111
          BC099414.1 - 1 - 2 148 30931867 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000239.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erich5

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144781 BC030476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144781 NM_173421.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173358 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127342 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109566 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144781 BC030476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144781 NM_173421.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30128634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03008027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109566 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08331529 - -
          Whole Mouse Genome G4122A A_51_P330825 BC030476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P330825 NM_173421.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129388 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08331529 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127342 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12151620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12151609 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12151613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30128751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12160565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12460966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12233445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12051128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12255460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12051121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12148579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12148566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11879550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12255459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30128628 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10423626 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423625 - -
          Affymetrix Mouse Gene 1.1 ST Array 10423623 - -
          Affymetrix Mouse Clariom S Array PSR1500001774.mm.1 NM_173421.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001774.mm.1 NM_173421.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001774.mm.1 TC1500000239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500001775.mm.1 NM_173421.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001775.mm.1 NM_173421.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001775.mm.1 TC1500000239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500001776.mm.1 NM_173421.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001776.mm.1 NM_173421.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500001776.mm.1 TC1500000239.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652378 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6380148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2597343 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2597343 - -
          mouse-6 v1.1 (Illumina) scl47999.3_178-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47999.3_178-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2597343 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2819380 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2819380 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2597343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0