Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558788_s1
          See other GPR82 GE Assays ›
          Gene Symbol:
          Gpr82
          Gene Name:
          G protein-coupled receptor 82
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 13658153 - 13667433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558788_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175669.3 NP_783600.1 4 730 105
          GenBank mRNA AK033761.1 - 4 732 105
          AY255571.1 - 1 65 105
          BC116704.1 - 4 725 40104936 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000171.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr82

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          biological_process
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044922 NM_175669.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044922 AK033761.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144192 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044922 NM_175669.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044922 AK033761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P166275 NM_175669.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P166275 AK033761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P166275 BC116704.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377771 NM_175669.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377771 AK033761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377771 AY255571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377771 BC116704.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32267432 - -
          Whole Mouse Genome G4122A A_52_P377771 NM_175669.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377771 AK033761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377771 AY255571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377771 BC116704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P166275 NM_175669.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P166275 AK033761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P166275 BC116704.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110605 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14916639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32267379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14662498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14690563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14690567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14662495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14916643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32267405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598762 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598757 - -
          Affymetrix Mouse Clariom S Array PSR0X00001114.mm.1 NM_175669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001114.mm.1 NM_175669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001114.mm.1 TC0X00000171.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496032 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2810524 - -
          Illumina mouse-6 v1.0 expression beadchip 2810524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238020 - -
          mouse-6 v1.1 (Illumina) scl55036.4_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl55036.4_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0