Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558811_g1
          See other TRIM12C GE Assays ›
          Gene Symbol:
          Trim12c
          Gene Name:
          tripartite motif-containing 12C
          Gene Aliases:
          9230105E10Rik, Trim12-2
          Chromosome Location:
          Chr.7: 104338754 - 104353358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558811_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim12c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146007.1 NP_001139479.1 7 - 8 1123 85
          NM_175677.4 NP_783608.3 7 - 8 1004 85
          GenBank mRNA AK033764.1 - 7 - 8 1124 85
          AK050485.1 - 7 - 8 1128 85
          AK146437.1 - 7 - 8 1083 85
          BC038937.1 - 7 - 8 1004 2648191 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003940.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim12c

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          protein ubiquitination
          ubiquitin-protein transferase activity
          signaling pattern recognition receptor activity
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163857 AK146437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163857 BC038937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163857 NM_175677.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163857 NM_001146007.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064652 BC062252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064652 BU923784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064652 AK050485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064652 AK146437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064652 BC038937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039061 AK146437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039061 BC038937.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039061 NM_175677.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039061 AK033764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039061 NM_001146007.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170083 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039061 AK146437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039061 BC038937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039061 NM_175677.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039061 AK033764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039061 NM_001146007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163857 AK146437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163857 BC038937.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163857 NM_175677.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163857 NM_001146007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064652 BC062252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064652 BU923784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064652 AK050485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064652 AK146437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064652 BC038937.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387834 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06308868 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01595099 - -
          Whole Mouse Genome G4122A A_52_P50063 AK146437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50063 BC038937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50063 NM_175677.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50063 AK033764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50063 NM_001146007.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06645594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06645593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06263650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566340 - -
          Affymetrix HT MG-430 PM Array Plate 1445271_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443858_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566333 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566346 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601581 - -
          Mouse Genome 430 2.0 Array 1445271_PM_at - -
          Mouse Genome 430 2.0 Array 1445271_at - -
          Affymetrix Mouse Clariom S Array PSR0700032931.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032931.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032931.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032931.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032931.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032927.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032927.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032927.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032927.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032927.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032932.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032932.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032932.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032932.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032932.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032929.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032929.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032929.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032929.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032929.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032923.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032923.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032923.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032923.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032923.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032924.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032924.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032924.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032924.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032924.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032930.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032930.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032930.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032930.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032930.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032933.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032933.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032933.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032922.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032922.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032922.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032922.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032922.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032926.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032926.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032926.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032926.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032926.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032925.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032925.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032925.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032925.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032925.mm.1 TC0700003940.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032934.mm.1 NM_001146007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032934.mm.1 NM_001146007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032934.mm.1 NM_175677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032934.mm.1 NM_175677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032934.mm.1 TC0700003940.mm.1 MTA TC
          Mouse Expression Set 430 1445271_at - -
          Mouse Expression Set 430 1445271_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445271_at - -
          Affymetrix HT Mouse Genome 430B Array 1445271_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 654536 - -
          ABI Mouse Genome Survey Microarray 720868 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5670041 - -
          MouseWG-6 v2 Expression BeadChip 6020593 - -
          Illumina mouse-6 v1.0 expression beadchip 6020593 - -
          Illumina mouse-6 v1.0 expression beadchip 5670041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661982 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661982 - -
          mouse-6 v1.1 (Illumina) scl0319236.1_50-S - -
          mouse-6 v1.1 (Illumina) scl00319236.1_170-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020593 - -
          Illumina Sentrix mouse-6.v1 bead array 5670041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319236.1_50-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319236.1_170-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020593 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661982 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline