Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558881_m1
          See other PLXNA4 GE Assays ›
          Gene Symbol:
          Plxna4
          Gene Name:
          plexin A4
          Gene Aliases:
          9330117B14, Plxa4, mKIAA1550
          Chromosome Location:
          Chr.6: 32144543 - 32588192 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558881_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plxna4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175750.3 NP_786926.2 2 - 3 2159 67
          GenBank mRNA AB073228.1 - 2 - 3 2158 67
          BC141298.1 - 2 - 3 1457 9055910 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002024.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plxna4

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          nervous system development
          axon guidance
          motor neuron axon guidance
          cranial nerve morphogenesis
          facial nerve morphogenesis
          facial nerve structural organization
          glossopharyngeal nerve morphogenesis
          trigeminal nerve morphogenesis
          trigeminal nerve structural organization
          vagus nerve morphogenesis
          postganglionic parasympathetic fiber development
          branchiomotor neuron axon guidance
          chemorepulsion of branchiomotor axon
          anterior commissure morphogenesis
          sympathetic nervous system development
          neuron projection morphogenesis
          regulation of axon extension involved in axon guidance
          regulation of negative chemotaxis
          semaphorin-plexin signaling pathway
          semaphorin-plexin signaling pathway involved in axon guidance
          protein binding
          semaphorin receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600015974.mm.1 TC0600002024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015974.mm.1 NM_175750.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015974.mm.1 NM_175750.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015977.mm.1 TC0600002024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015977.mm.1 NM_175750.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015977.mm.1 NM_175750.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015975.mm.1 TC0600002024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015975.mm.1 NM_175750.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015975.mm.1 NM_175750.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015978.mm.1 TC0600002024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015978.mm.1 NM_175750.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015978.mm.1 NM_175750.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015976.mm.1 TC0600002024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015976.mm.1 NM_175750.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015976.mm.1 NM_175750.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline