Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00613004_m1
          See other EMC3 GE Assays ›
          Gene Symbol:
          Emc3
          Gene Name:
          ER membrane protein complex subunit 3
          Gene Aliases:
          0610039A15Rik, AI225901, AW260416, Pob, Tmem111
          Chromosome Location:
          Chr.6: 113514887 - 113531638 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00613004_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175101.3 NP_780310.1 3 - 4 463 102
          GenBank mRNA AK020058.1 - 3 - 4 434 102
          AK075595.1 - 3 - 4 464 102
          AK167819.1 - 3 - 4 312 102
          AK206599.1 - 3 - 4 209 102
          AK213319.1 - 3 - 4 210 103
          AK218418.1 - 3 - 4 210 103
          BC004641.1 - 3 - 4 459 3157147 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002924.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emc3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein folding in endoplasmic reticulum
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24760052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24760053 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 BC004641.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK167819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK020058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK075595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 NM_175101.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 BC004641.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK167819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK020058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 AK075595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56658 NM_175101.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426140 AK081344.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426140 BC052153.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426140 K02582.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426140 L10076.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426140 NM_021600.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117598 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121558 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157471 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 BC004641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK167819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK020058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK075595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 NM_175101.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 BC004641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK167819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK020058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 AK075595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56658 NM_175101.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24760033 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01421290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06031268 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06031258 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121558 - -
          Whole Mouse Genome G4122A A_52_P989682 - -
          Whole Mouse Genome G4122A A_52_P56658 BC004641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK167819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK020058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK075595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 NM_175101.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 BC004641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK167819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK020058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 AK075595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56658 NM_175101.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112719 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01421290 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157471 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05297197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05297193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24760053 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24760052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24760024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05538957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05371251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05426848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05096262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05538960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24760007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546950 - -
          Affymetrix HT MG-430 PM Array Plate 1451145_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10546944 - -
          Mouse Genome 430 2.0 Array 1451145_PM_s_at - -
          Mouse Genome 430 2.0 Array 1451145_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66087_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66087_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600022475.mm.1 TC0600002924.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022475.mm.1 NM_175101.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022475.mm.1 NM_175101.3 Refseq
          Mouse Expression Set 430 1451145_PM_s_at - -
          Mouse Expression Set 430 1451145_s_at - -
          Murine Genome U74v2 Set 97862_PM_s_at - -
          Murine Genome U74v2 Set 97861_PM_at - -
          Murine Genome U74v2 Set 97861_at - -
          Murine Genome U74v2 Set 97862_s_at - -
          Mouse Genome 430A 2.0 Array 1451145_s_at - -
          Mouse Genome 430A 2.0 Array 1451145_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451145_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451145_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 583439 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770594 - -
          MouseWG-6 v2 Expression BeadChip 5720441 - -
          Illumina mouse-6 v1.0 expression beadchip 5720441 - -
          Illumina mouse-6 v1.0 expression beadchip 6770594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689083 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689083 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228253 - -
          mouse-6 v1.1 (Illumina) scl28539.8_61-S - -
          mouse-6 v1.1 (Illumina) scl0001142.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720441 - -
          Illumina Sentrix mouse-6.v1 bead array 6770594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001142.1_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28539.8_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2859294 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689083 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228253 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689083 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2859294 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228253 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001142.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28539.8_61-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline