Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617772_g1
          See other GALE GE Assays ›
          Gene Symbol:
          Gale
          Gene Name:
          galactose-4-epimerase, UDP
          Gene Aliases:
          2310002A12Rik, AI323962
          Chromosome Location:
          Chr.4: 135963681 - 135968178 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617772_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gale
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178389.3 NP_848476.1 4 - 5 698 93
          GenBank mRNA AK009083.1 - 5 - 6 565 93
          AK146184.1 - 5 - 6 590 93
          BC027438.1 - 4 - 5 698 4224021 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001671.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gale

          Panther Classification:

          Molecular Function -

          epimerase/racemase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          galactose metabolic process
          galactose catabolic process
          catalytic activity
          UDP-N-acetylglucosamine 4-epimerase activity
          UDP-glucose 4-epimerase activity
          isomerase activity
          racemase and epimerase activity, acting on carbohydrates and derivatives
          protein homodimerization activity
          coenzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331442 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111855 AK146184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111855 BC027438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111855 NM_178389.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733207 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114941 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111855 AK146184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111855 BC027438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111855 NM_178389.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401193 AK009083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401193 AK146184.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401193 BC027438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401193 NM_178389.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23331442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00997256 - -
          Whole Mouse Genome G4122A A_51_P401193 AK009083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401193 AK146184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401193 BC027438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401193 NM_178389.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129538 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101173 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03697309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03697308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03302199 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03302184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03302200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03302182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23331445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23331515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03697307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03302188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23331471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509155 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509160 - -
          Affymetrix HT MG-430 PM Array Plate 1424140_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509147 - -
          Mouse Genome 430 2.0 Array 1424140_PM_at - -
          Mouse Genome 430 2.0 Array 1424140_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74246_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74246_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400013399.mm.1 NM_178389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013399.mm.1 NM_178389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013399.mm.1 TC0400001671.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013398.mm.1 NM_178389.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013398.mm.1 NM_178389.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013398.mm.1 TC0400001671.mm.1 MTA TC
          Mouse Expression Set 430 1424140_PM_at - -
          Mouse Expression Set 430 1424140_at - -
          Murine Genome U74v2 Set 93566_at - -
          Murine Genome U74v2 Set 93566_PM_at - -
          Mouse Genome 430A 2.0 Array 1424140_at - -
          Mouse Genome 430A 2.0 Array 1424140_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424140_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424140_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 859085 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290072 - -
          MouseWG-6 v2 Expression BeadChip 3610400 - -
          Illumina mouse-6 v1.0 expression beadchip 3610400 - -
          Illumina mouse-6 v1.0 expression beadchip 6290072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2683811 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237990 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683811 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237990 - -
          mouse-6 v1.1 (Illumina) scl074246.9_1-S - -
          mouse-6 v1.1 (Illumina) scl24826.5.1_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610400 - -
          Illumina Sentrix mouse-6.v1 bead array 6290072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl074246.9_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24826.5.1_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237990 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2683811 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2683811 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline