Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617930_m1
          See other CHIL6 GE Assays ›
          Gene Symbol:
          Chil6
          Gene Name:
          chitinase-like 6
          Gene Aliases:
          BYm, Chil8
          Chromosome Location:
          Chr.3: 106387384 - 106409632 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chil6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178412.2 NP_848499.1 10 - 11 1378 82
          GenBank mRNA BC051070.1 - 10 - 11 1378 6308125 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002686.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chil6

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          chitin catabolic process
          biological_process
          molecular_function
          chitinase activity
          chitin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004162 NM_178412.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004162 BC051070.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62122 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106911 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151400 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116755 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004162 NM_178412.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004162 BC051070.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22401671 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106911 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116755 - -
          Whole Mouse Genome G4122A A_51_P456376 NM_178412.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456376 BC051070.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130278 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104542 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116755 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151400 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02574697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03010133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03010132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03010125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22401675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02574704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22401697 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10501035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501041 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501043 - -
          Affymetrix Mouse Gene 1.1 ST Array 10501034 - -
          Affymetrix Mouse Clariom S Array PSR0300020935.mm.1 NM_178412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020935.mm.1 NM_178412.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020935.mm.1 TC0300002686.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 816874 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4280019 - -
          Illumina mouse-6 v1.0 expression beadchip 4280019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231143 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231143 - -
          mouse-6 v1.1 (Illumina) scl21697.14.1_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21697.14.1_152-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231143 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline