Search
Search

Commd8| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178599.4 | NP_848714.1 | 2 - 3 | 261 | 78 | |
| GenBank mRNA | AK009275.1 | - | 2 - 3 | 250 | 78 | |
| AK012653.1 | - | 2 - 3 | 247 | 78 | ||
| AK013371.1 | - | 2 - 3 | 234 | 78 | ||
| AK077934.1 | - | 2 - 3 | 261 | 78 | ||
| AK151560.1 | - | 2 - 3 | 244 | 78 | ||
| BC098183.1 | - | 2 - 3 | 237 | 5150466 | 78 | |
Set Membership: |
|