Search
Search

Wdr24| Gene Symbol: Wdr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173741.3 | NP_776102.1 | 7 - 8 | 2597 | 65 | |
| GenBank mRNA | AK078694.1 | - | 7 - 8 | 2578 | 65 | |
| AK170253.1 | - | 6 - 7 | 2648 | 65 | ||
| BC037651.1 | - | 7 - 8 | 2519 | 5148210 | 65 | |
Set Membership: |
|