Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620605_m1
          See other SLC45A1 GE Assays ›
          Gene Symbol:
          Slc45a1
          Gene Name:
          solute carrier family 45, member 1
          Gene Aliases:
          C230078B22, Dnb5
          Chromosome Location:
          Chr.4: 150629396 - 150652174 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620605_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc45a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173774.3 NP_776135.2 5 - 6 1630 78
          GenBank mRNA AK082651.1 - 1623 78
          AK134677.1 - 5 - 6 1556 78
          AK134683.1 - 5 - 6 1621 78
          BC051188.1 - 5 - 6 1592 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004066.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc45a1

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          carbohydrate transport
          glucose transport
          hexose transmembrane transport
          transmembrane transport
          glucose transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407528 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081785 BC051188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081785 BM934007.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081785 AK134683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081785 NM_173774.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081785 AK082651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071888 AK134677.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131086 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157598 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110929 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081785 BC051188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081785 BM934007.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081785 AK134683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081785 NM_173774.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081785 AK082651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071888 AK134677.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281486 AK082651.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23407522 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05450278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110929 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01017925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157598 - -
          Whole Mouse Genome G4122A A_51_P281486 AK082651.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102555 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157598 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03435106 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03435110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03959081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03959080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03435105 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23407528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23407398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03508598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03936623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03220119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03965263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03692955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03959078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03550590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03845341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03591129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03959080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03352695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03591130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03352694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03508600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03550589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03781076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03991831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03995933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03936621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03910972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03508599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03529397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03508602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03435110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03851558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03959081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23407560 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518768 - -
          Affymetrix HT MG-430 PM Array Plate 1436252_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518763 - -
          Mouse Genome 430 2.0 Array 1436252_at - -
          Mouse Genome 430 2.0 Array 1436252_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242773_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242773_at - -
          Affymetrix Mouse Clariom S Array PSR0400033794.mm.1 TC0400004066.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033794.mm.1 NM_173774.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033794.mm.1 NM_173774.3 Refseq
          Mouse Expression Set 430 1436252_at - -
          Mouse Expression Set 430 1436252_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436252_at - -
          Affymetrix HT Mouse Genome 430B Array 1436252_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 464918 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3780162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163567 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242488 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242488 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163567 - -
          mouse-6 v1.1 (Illumina) scl23481.9_162-S - -
          mouse-6 v1.1 (Illumina) GI_75709221-A - -
          Illumina Sentrix mouse-6.v1 bead array 3780162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23481.9_162-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163567 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242488 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163567 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline