Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00624191_m1
          See other SCGB2B24 GE Assays ›
          Gene Symbol:
          Scgb2b24
          Gene Name:
          secretoglobin, family 2B, member 24
          Gene Aliases:
          Abpbg24, Abpz, C2b, Scgb2b3
          Chromosome Location:
          Chr.7: 33737193 - 33739295 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00624191_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb2b24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177446.2 NP_803229.1 2 - 3 256 101
          GenBank mRNA AA794526.1 - 2 - 3 256 101
          AY370635.1 - 2 - 3 273 101
          BC115602.1 - 2 - 3 256 40102624 101
          BC115603.1 - 2 - 3 256 40102625 101
          GU269236.1 - 2 - 3 246 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002816.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb2b24

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002050 NM_177446.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002050 AY370635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002050 BC115602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002050 BC115603.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002050 GU269236.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112003 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120665 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002050 NM_177446.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002050 AY370635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002050 BC115602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002050 BC115603.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002050 GU269236.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25041312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120665 - -
          Whole Mouse Genome G4122A A_52_P570679 NM_177446.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570679 AY370635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570679 BC115602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570679 BC115603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P570679 GU269236.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110129 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05911328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25041303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05911328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05911323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05911316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05911308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06493981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25041307 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562303 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562299 - -
          Affymetrix Mouse Clariom S Array PSR0700025167.mm.1 TC0700002816.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025167.mm.1 NM_177446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025167.mm.1 NM_177446.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025168.mm.1 TC0700002816.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025168.mm.1 NM_177446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025168.mm.1 NM_177446.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025166.mm.1 TC0700002816.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025166.mm.1 NM_177446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025166.mm.1 NM_177446.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025169.mm.1 TC0700002816.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025169.mm.1 NM_177446.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025169.mm.1 NM_177446.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 409555 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232401 - -
          mouse-6 v1.1 (Illumina) scl31492.3.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31492.3.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232401 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline