Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00624380_m1
          See other SLFNL1 GE Assays ›
          Gene Symbol:
          Slfnl1
          Gene Name:
          schlafen like 1
          Gene Aliases:
          4933406A14Rik
          Chromosome Location:
          Chr.4: 120531981 - 120536661 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00624380_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slfnl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177570.3 NP_808238.1 2 - 3 827 149
          GenBank mRNA AK077106.1 - 2 - 3 828 149
          BC118052.1 - 1 - 2 578 40105639 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001352.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slfnl1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          molecular_function
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 BC117545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 BC118052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 AK077106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 NM_177570.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 BC117545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 BC118052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 AK077106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P149687 NM_177570.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 650449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123660 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 BC117545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 BC118052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 AK077106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 NM_177570.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 BC117545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 BC118052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 AK077106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P149687 NM_177570.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23251743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168421 - -
          Whole Mouse Genome G4122A A_52_P149687 BC117545.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 BC118052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 AK077106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 NM_177570.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 BC117545.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 BC118052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 AK077106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149687 NM_177570.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128571 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03280867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03280886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03280894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03280896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03280887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23251784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03614325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03614349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03614327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23251770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507724 - -
          Affymetrix HT MG-430 PM Array Plate 1455838_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10507719 - -
          Mouse Genome 430 2.0 Array 1455838_at - -
          Mouse Genome 430 2.0 Array 1455838_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194219_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194219_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400010282.mm.1 NM_177570.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010282.mm.1 NM_177570.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400010282.mm.1 TC0400001352.mm.1 MTA TC
          Mouse Expression Set 430 1455838_at - -
          Mouse Expression Set 430 1455838_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455838_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455838_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 404460 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320750 - -
          Illumina mouse-6 v1.0 expression beadchip 2320750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245376 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245376 - -
          mouse-6 v1.1 (Illumina) scl25014.5.1_128-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25014.5.1_128-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245376 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245376 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline