Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00624399_s1
          See other DOLK GE Assays ›
          Gene Symbol:
          Dolk
          Gene Name:
          dolichol kinase
          Gene Aliases:
          BC026973, Tmem15, mKIAA1094
          Chromosome Location:
          Chr.2: 30284229 - 30286354 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00624399_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dolk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177648.3 NP_808316.1 1 1622 57
          GenBank mRNA AB073968.1 - 1 1565 57
          AK077581.1 - 1 1593 57
          AK144705.1 - 1 1603 57
          BC019543.1 - 1 1567 57
          BC026973.1 - 1 1347 5371267 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dolk

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorylation
          dolichyl monophosphate biosynthetic process
          dolichol kinase activity
          kinase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102297 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 BC026973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK144705.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK214790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK217764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 BC019543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 BC026973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK144705.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK214790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 AK217764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284966 BC019543.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785127 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179473 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131001 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133514 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 BC026973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK144705.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK214790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK217764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 BC019543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 BC026973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK144705.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK214790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 AK217764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284966 BC019543.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21102295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131001 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179473 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133514 - -
          Whole Mouse Genome G4122A A_51_P284966 BC026973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK144705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK214790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK217764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 BC019543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 BC026973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK144705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK214790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 AK217764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284966 BC019543.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118948 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179473 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01335426 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01335448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01335447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01335439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21102295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21102286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01873571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01238098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21102291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01873578 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481454 - -
          Affymetrix HT MG-430 PM Array Plate 1424366_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481453 - -
          Mouse Genome 430 2.0 Array 1424366_PM_at - -
          Mouse Genome 430 2.0 Array 1424366_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227697_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227697_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200027612.mm.1 NM_177648.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027612.mm.1 NM_177648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027612.mm.1 TC0200003267.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027613.mm.1 NM_177648.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027613.mm.1 NM_177648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027613.mm.1 TC0200003267.mm.1 MTA TC
          Mouse Expression Set 430 1424366_at - -
          Mouse Expression Set 430 1424366_PM_at - -
          Murine Genome U74v2 Set 166119_at - -
          Murine Genome U74v2 Set 166119_PM_at - -
          Mouse Genome 430A 2.0 Array 1424366_PM_at - -
          Mouse Genome 430A 2.0 Array 1424366_at - -
          Affymetrix HT Mouse Genome 430A Array 1424366_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424366_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 577485 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220524 - -
          Illumina mouse-6 v1.0 expression beadchip 5220524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226126 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226126 - -
          mouse-6 v1.1 (Illumina) scl19473.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19473.1_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226126 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226126 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19473.1_35-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline