Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626457_m1
          See other PRAMEL6 GE Assays ›
          Gene Symbol:
          Pramel6
          Gene Name:
          preferentially expressed antigen in melanoma like 6
          Gene Aliases:
          C80453, C80515
          Chromosome Location:
          Chr.2: 87506565 - 87510868 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626457_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pramel6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178249.2 NP_839980.1 2 - 3 873 115
          GenBank mRNA AF490342.1 - 2 - 3 873 115
          AK139830.1 - 3 - 4 1107 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001294.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pramel6

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161590 AK139830.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26354 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169192 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161590 AK139830.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00425705 - -
          Whole Mouse Genome G4122A A_52_P455379 NM_178249.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455379 AF490342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455379 AK139830.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120120 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377456 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01377475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01673416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01650382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01726678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01658762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01377460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01377439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01733379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01450549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01916302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01156660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01377471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02003126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02110536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01673413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473544 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473545 - -
          Affymetrix HT MG-430 PM Array Plate 1456269_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456270_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473543 - -
          Mouse Genome 430 2.0 Array 1456269_at - -
          Mouse Genome 430 2.0 Array 1456269_PM_at - -
          Mouse Genome 430 2.0 Array 1456270_s_at - -
          Mouse Genome 430 2.0 Array 1456270_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 347711_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 347711_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200010056.mm.1 NM_178249.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010056.mm.1 NM_178249.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010056.mm.1 TC0200001294.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010055.mm.1 NM_178249.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010055.mm.1 NM_178249.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010055.mm.1 TC0200001294.mm.1 MTA TC
          Mouse Expression Set 430 1456270_s_at - -
          Mouse Expression Set 430 1456269_PM_at - -
          Mouse Expression Set 430 1456270_PM_s_at - -
          Mouse Expression Set 430 1456269_at - -
          Murine Genome U74v2 Set 103470_PM_at - -
          Murine Genome U74v2 Set 103470_at - -
          Mouse Genome 430A 2.0 Array 1456269_at - -
          Mouse Genome 430A 2.0 Array 1456270_s_at - -
          Mouse Genome 430A 2.0 Array 1456269_PM_at - -
          Mouse Genome 430A 2.0 Array 1456270_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1456270_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1456270_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1456269_at - -
          Affymetrix HT Mouse Genome 430A Array 1456269_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 590714 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2370593 - -
          Illumina mouse-6 v1.0 expression beadchip 2370593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228899 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228899 - -
          mouse-6 v1.1 (Illumina) scl20672.6.1_211-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20672.6.1_211-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228899 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228899 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline