Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652120_mH
          See other MS4A4C GE Assays ›
          Gene Symbol:
          Ms4a4c
          Gene Name:
          membrane-spanning 4-domains, subfamily A, member 4C
          Gene Aliases:
          5830413L19Rik, AI449185, Ms4a9
          Chromosome Location:
          Chr.19: 11404717 - 11427246 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652120_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ms4a4c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029499.3 NP_083775.1 2 - 3 268 144
          GenBank mRNA AF237914.2 - 2 - 3 242 144
          AK017928.1 - 2 - 3 248 144
          AK151045.1 - 2 - 3 261 144
          AK153495.1 - 1 - 2 112 144
          AK157137.1 - 270 144
          AK172650.1 - 2 - 3 240 144
          BC027496.1 - 2 - 3 224 1383416 144
          CT010170.1 - 1 - 2 136 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000217.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ms4a4c

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135986 AK157137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135986 AK172650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135986 BC027496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135986 CT010170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135986 AF237914.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106388 AK172650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106388 BC027496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106388 AF237914.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106388 AK017928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106388 AK151045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135980 AK172650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135980 BC027496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135980 AF237914.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135980 AK017928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135980 AK151045.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147159 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122342 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109344 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135986 AK157137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135986 AK172650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135986 BC027496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135986 CT010170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135986 AF237914.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106388 AK172650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106388 BC027496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106388 AF237914.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106388 AK017928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106388 AK151045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135980 AK172650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135980 BC027496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135980 AF237914.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135980 AK017928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135980 AK151045.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955942 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09106341 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03550565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122342 - -
          Whole Mouse Genome G4122A A_52_P393488 AK157137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393488 AK172650.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393488 BC027496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393488 CT010170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393488 AF237914.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302977 AB026045.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106388 AK172650.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106388 BC027496.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106388 AF237914.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106388 AK017928.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106388 AK151045.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140788 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109344 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14299285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14226567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14226565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14264403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14226567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14173534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14487232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14173529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31955951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461600 - -
          Affymetrix HT MG-430 PM Array Plate 1420671_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1450291_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461594 - -
          Mouse Genome 430 2.0 Array 1420671_PM_x_at - -
          Mouse Genome 430 2.0 Array 1450291_PM_s_at - -
          Mouse Genome 430 2.0 Array 1450291_s_at - -
          Mouse Genome 430 2.0 Array 1420671_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64380_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64380_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002706.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002706.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002706.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002707.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002707.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002707.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002716.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002716.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002716.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002708.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002708.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002708.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002713.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002713.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002713.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002715.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002715.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002715.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002711.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002711.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002711.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002712.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002712.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002712.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002714.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002714.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002714.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002704.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002704.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002704.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002710.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002710.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002710.mm.1 TC1900000217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002709.mm.1 NM_029499.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002709.mm.1 NM_029499.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002709.mm.1 TC1900000217.mm.1 MTA TC
          Mouse Expression Set 430 1450291_s_at - -
          Mouse Expression Set 430 1420671_x_at - -
          Mouse Expression Set 430 1450291_PM_s_at - -
          Mouse Expression Set 430 1420671_PM_x_at - -
          Murine Genome U74v2 Set 98373_PM_at - -
          Murine Genome U74v2 Set 98373_at - -
          Mouse Genome 430A 2.0 Array 1450291_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1420671_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1420671_x_at - -
          Mouse Genome 430A 2.0 Array 1450291_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450291_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420671_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420671_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450291_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 495283 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360605 - -
          Illumina mouse-6 v1.0 expression beadchip 5360605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2678607 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2678607 - -
          mouse-6 v1.1 (Illumina) scl064380.6_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360605 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl064380.6_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2678607 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2678607 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline