Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652250_s1
          See other VMN1R46 GE Assays ›
          Gene Symbol:
          Vmn1r46
          Gene Name:
          vomeronasal 1 receptor 46
          Gene Aliases:
          V1rb8
          Chromosome Location:
          Chr.6: 89969304 - 89977100 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652250_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053229.1 NP_444459.1 1 439 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r46

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          response to pheromone
          signal transducer activity
          receptor activity
          G-protein coupled receptor activity
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014801 NM_053229.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111690 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014801 NM_053229.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469724 NM_053229.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24630485 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111690 - -
          Whole Mouse Genome G4122A A_51_P469724 NM_053229.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133634 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05587546 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05587545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05587548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05587534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05587550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24630463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05411979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05587545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05414354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24630479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539964 - -
          Affymetrix HT MG-430 PM Array Plate 1450601_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539963 - -
          Mouse Genome 430 2.0 Array 1450601_at - -
          Mouse Genome 430 2.0 Array 1450601_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113856_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113856_PM_at - -
          Mouse Expression Set 430 1450601_at - -
          Mouse Expression Set 430 1450601_PM_at - -
          Mouse Genome 430A 2.0 Array 1450601_PM_at - -
          Mouse Genome 430A 2.0 Array 1450601_at - -
          Affymetrix HT Mouse Genome 430A Array 1450601_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450601_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332470 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2760577 - -
          Illumina mouse-6 v1.0 expression beadchip 102190164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230115 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230115 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213719 - -
          mouse-6 v1.1 (Illumina) GI_16716540-S - -
          mouse-6 v1.1 (Illumina) scl0113856.1_226-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760577 - -
          Illumina Sentrix mouse-6.v1 bead array 102190164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0113856.1_226-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760577 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102190164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230115 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230115 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline