Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652746_s1
          See other VMN1R75 GE Assays ›
          Gene Symbol:
          Vmn1r75
          Gene Name:
          vomeronasal 1 receptor 75
          Gene Aliases:
          V1rg6
          Chromosome Location:
          Chr.7: 11880343 - 11881260 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652746_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134207.1 NP_598968.1 1 351 129
          GenBank mRNA BC115506.1 - 1 344 129
          BC115507.1 - 1 351 40101051 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r75

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 NM_134207.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 BC115506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 NM_134207.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 BC115506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169274 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 NM_134207.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 BC115506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 BC115507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 NM_134207.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 BC115506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 NM_134207.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 BC115506.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 NM_134207.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 BC115506.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969951 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169274 - -
          Whole Mouse Genome G4122A A_51_P137742 NM_134207.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137742 BC115506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137742 BC115507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137742 NM_134207.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137742 BC115506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137742 BC115507.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114127 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06343375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06343373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06343362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05929906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06343364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06514758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05929905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05945325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06343362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06647424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05888908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05945331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06163000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05945326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969916 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549987 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549986 - -
          Affymetrix Mouse Clariom S Array PSR0700001033.mm.1 TC0700000122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001033.mm.1 NM_134207.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001033.mm.1 NM_134207.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 871904 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190309 - -
          MouseWG-6 v2 Expression BeadChip 106020398 - -
          Illumina mouse-6 v1.0 expression beadchip 106020398 - -
          Illumina mouse-6 v1.0 expression beadchip 2190309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245439 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241654 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245439 - -
          mouse-6 v1.1 (Illumina) GI_21717728-S - -
          mouse-6 v1.1 (Illumina) scl9722.1.1_99-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190309 - -
          Illumina Sentrix mouse-6.v1 bead array 106020398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9722.1.1_99-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190309 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106020398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245439 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline