Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653044_m1
          See other MCHR1 GE Assays ›
          Gene Symbol:
          Mchr1
          Gene Name:
          melanin-concentrating hormone receptor 1
          Gene Aliases:
          AW049955, Gpr24, Mch1r
          Chromosome Location:
          Chr.15: 81235499 - 81238964 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653044_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mchr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145132.2 NP_660114.1 1 - 2 357 83
          GenBank mRNA AF498247.1 - 1 - 2 82 83
          AF498248.1 - 1 - 2 82 83
          AK134867.1 - 1 - 2 240 83
          BC128285.1 - 1 - 2 82 83
          BC128286.1 - 1 - 2 82 40102363 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mchr1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          positive regulation of calcium ion transport
          regulation of feeding behavior
          signal transducer activity
          G-protein coupled receptor activity
          protein C-terminus binding
          melanin-concentrating hormone receptor activity
          hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021146 AF498248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021146 AK134867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021146 BC128285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021146 BC128286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021146 NM_145132.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798444 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104771 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021146 AF498248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021146 AK134867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021146 BC128285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021146 BC128286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021146 NM_145132.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376299 AF498248.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376299 AK134867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376299 BC128285.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376299 BC128286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376299 NM_145132.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30376903 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104771 - -
          Whole Mouse Genome G4122A A_51_P376299 AF498248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376299 AK134867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376299 BC128285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376299 BC128286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376299 NM_145132.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124099 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421433 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12421438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30376901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12008327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11920544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12062690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11921914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12421433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12253985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12253981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12421448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11905383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12358225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12133227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11961993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12010888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12341278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30376885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11921913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12421439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425509 - -
          Affymetrix HT MG-430 PM Array Plate 1454844_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454845_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1428031_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425507 - -
          Mouse Genome 430 2.0 Array 1454844_PM_at - -
          Mouse Genome 430 2.0 Array 1428031_at - -
          Mouse Genome 430 2.0 Array 1454845_x_at - -
          Mouse Genome 430 2.0 Array 1454844_at - -
          Mouse Genome 430 2.0 Array 1428031_PM_at - -
          Mouse Genome 430 2.0 Array 1454845_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207911_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207911_at - -
          Affymetrix Mouse Clariom S Array PSR1500005477.mm.1 NM_145132.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005477.mm.1 NM_145132.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005477.mm.1 TC1500000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005479.mm.1 NM_145132.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005479.mm.1 NM_145132.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005479.mm.1 TC1500000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005478.mm.1 NM_145132.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005478.mm.1 NM_145132.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005478.mm.1 TC1500000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005480.mm.1 NM_145132.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005480.mm.1 NM_145132.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005480.mm.1 TC1500000718.mm.1 MTA TC
          Mouse Expression Set 430 1428031_PM_at - -
          Mouse Expression Set 430 1428031_at - -
          Mouse Expression Set 430 1454845_PM_x_at - -
          Mouse Expression Set 430 1454844_at - -
          Mouse Expression Set 430 1454845_x_at - -
          Mouse Expression Set 430 1454844_PM_at - -
          Mouse Genome 430A 2.0 Array 1428031_PM_at - -
          Mouse Genome 430A 2.0 Array 1428031_at - -
          Affymetrix HT Mouse Genome 430A Array 1428031_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428031_at - -
          Affymetrix HT Mouse Genome 430B Array 1454845_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1454844_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454844_at - -
          Affymetrix HT Mouse Genome 430B Array 1454845_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 684785 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603364 - -
          mouse-6 v1.1 (Illumina) scl47713.3_671-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47713.3_671-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2603364 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2603364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline