Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654584_s1
          See other OLFR1301 GE Assays ›
          Gene Symbol:
          Olfr1301
          Gene Name:
          olfactory receptor 1301
          Gene Aliases:
          MOR248-5
          Chromosome Location:
          Chr.2: 111751132 - 111755189 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654584_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1301
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146887.1 NP_667098.1 1 161 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1301

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953201 NM_146887.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179940 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953201 NM_146887.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P303296 NM_146887.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21526351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179940 - -
          Whole Mouse Genome G4122A A_51_P303296 NM_146887.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134750 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01507134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01507139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01507122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01507116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21526397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02104450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02104442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21526392 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474515 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474514 - -
          Affymetrix Mouse Clariom S Array PSR0200012302.mm.1 NM_146887.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012302.mm.1 NM_146887.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012302.mm.1 TC0200001654.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 804144 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270148 - -
          Illumina mouse-6 v1.0 expression beadchip 5270148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590984 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590984 - -
          mouse-6 v1.1 (Illumina) scl20491.1.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20491.1.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2590984 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline