Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655086_s1
          See other OLFR1047 GE Assays ›
          Gene Symbol:
          Olfr1047
          Gene Name:
          olfactory receptor 1047
          Gene Aliases:
          MOR188-3
          Chromosome Location:
          Chr.2: 86228010 - 86228969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655086_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1047
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147012.1 NP_667223.1 1 347 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1047

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375458 NM_147012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375458 NM_147012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375458 NM_147012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375458 NM_147012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375458 NM_147012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375458 NM_147012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375458 NM_147012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375458 NM_147012.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01783003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01783000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02184629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401386 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484574 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484573 - -
          Affymetrix Mouse Clariom S Array PSR0200033741.mm.1 NM_147012.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033741.mm.1 NM_147012.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033741.mm.1 TC0200003922.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 521142 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3710324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716006 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716006 - -
          mouse-6 v1.1 (Illumina) scl4344.1.1_213-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4344.1.1_213-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2716006 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2716006 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline