Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655373_m1
          See other UGT2B34 GE Assays ›
          Gene Symbol:
          Ugt2b34
          Gene Name:
          UDP glucuronosyltransferase 2 family, polypeptide B34
          Gene Aliases:
          AI788959
          Chromosome Location:
          Chr.5: 86889770 - 86906937 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655373_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt2b34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153598.2 NP_705826.1 5 - 6 1337 113
          GenBank mRNA BC028826.1 - 5 - 6 1337 5098890 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt2b34

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          flavonoid biosynthetic process
          cellular response to hormone stimulus
          flavonoid glucuronidation
          molecular_function
          glucuronosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977418 BC028826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977418 NM_153598.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977418 AK145951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977418 AK149413.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55924 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126484 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977418 BC028826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977418 NM_153598.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977418 AK145951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977418 AK149413.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432924 BC028826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432924 NM_153598.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432924 AK145951.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23865921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05643941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126484 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05643924 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118807 - -
          Whole Mouse Genome G4122A A_51_P432924 BC028826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432924 NM_153598.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432924 AK145951.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116953 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126484 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04245819 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04672023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04672017 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04245815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04245813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04672026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04245818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23865845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04337265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04231750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04245823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04245814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04337262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04337259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04245818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23865917 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531038 - -
          Affymetrix HT MG-430 PM Array Plate 1427961_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1427960_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531034 - -
          Mouse Genome 430 2.0 Array 1427961_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427960_PM_at - -
          Mouse Genome 430 2.0 Array 1427961_s_at - -
          Mouse Genome 430 2.0 Array 1427960_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100727_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100727_at - -
          Affymetrix Mouse Clariom S Array PSR0500023736.mm.1 TC0500002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023736.mm.1 NM_153598.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023736.mm.1 NM_153598.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023738.mm.1 TC0500002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023738.mm.1 NM_153598.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023738.mm.1 NM_153598.2 Refseq
          Murine Genome U74v2 Set 103227_at - -
          Murine Genome U74v2 Set 103227_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427960_at - -
          Affymetrix HT Mouse Genome 430A Array 1427961_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427961_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427960_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 612894 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980347 - -
          Illumina mouse-6 v1.0 expression beadchip 1980347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746475 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746475 - -
          mouse-6 v1.1 (Illumina) scl26423.7_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26423.7_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2746475 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746475 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline