Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00657594_m1
          See other ATF1 GE Assays ›
          Gene Symbol:
          Atf1
          Gene Name:
          activating transcription factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 100227859 - 100261248 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00657594_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007497.3 NP_031523.3 2 - 3 328 153
          GenBank mRNA AK010428.1 - 2 - 3 209 153
          AK144511.1 - 2 - 3 370 153
          BC006871.1 - 2 - 3 274 2646620 153
          BY733806.1 - 2 - 3 367 153
          M63725.1 - 1 - 2 118 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of neuron projection development
          cellular protein complex assembly
          positive regulation of DNA replication
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein complex binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500008199.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008199.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008199.mm.1 TC1500001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008200.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008200.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008200.mm.1 TC1500001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008214.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008214.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008214.mm.1 TC1500001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008215.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008215.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008215.mm.1 TC1500001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008213.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008213.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008213.mm.1 TC1500001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008209.mm.1 NM_007497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008209.mm.1 NM_007497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008209.mm.1 TC1500001038.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline