Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660275_m1
          See other NAGPA GE Assays ›
          Gene Symbol:
          Nagpa
          Gene Name:
          N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase
          Gene Aliases:
          AI596180, UCE
          Chromosome Location:
          Chr.16: 5195280 - 5204040 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660275_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nagpa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013796.3 NP_038824.2 2 - 3 571 61
          NR_027804.1 1 - 2 668 61
          GenBank mRNA AK032158.1 - 2 - 3 572 61
          AK087468.1 - 2 - 3 571 61
          AK138035.1 - 2 - 3 564 61
          AK155450.1 - 2 - 3 550 61
          AK159629.1 - 2 - 3 550 61
          AK162633.1 - 2 - 3 553 61
          AK168830.1 - 1 - 2 658 61
          AK170219.1 - 2 - 3 553 61
          BC019640.1 - 1 - 2 633 61
          BC039790.1 - 2 - 3 563 4951805 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001155.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nagpa

          Gene Ontology Categories:

          Process(es)

          N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514372 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK162633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK168830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK159629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK138035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK087468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK162633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK168830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK159629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK138035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P370050 AK087468.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148868 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124659 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115112 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128753 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK162633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK168830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK159629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK138035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK087468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK162633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK168830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK159629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK138035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P370050 AK087468.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30514385 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115112 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124659 - -
          Whole Mouse Genome G4122A A_51_P370050 AK162633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK168830.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK159629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK138035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK087468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK162633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK168830.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK159629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK138035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370050 AK087468.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121382 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124659 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12555616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12608567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12608580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30514374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30514297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12730186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12704931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13027709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12577126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12608572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12846738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12877936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12522740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12963651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12673196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13041330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12963653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12622751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12608584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12673195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30514408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12815797 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10437545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437547 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10437544 - -
          Affymetrix HT MG-430 PM Array Plate 1454718_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458344_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10437541 - -
          Mouse Genome 430 2.0 Array 1454718_PM_at - -
          Mouse Genome 430 2.0 Array 1458344_PM_at - -
          Mouse Genome 430 2.0 Array 1458344_at - -
          Mouse Genome 430 2.0 Array 1454718_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27426_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27426_at - -
          Affymetrix Mouse Clariom S Array PSR1600009261.mm.1 NM_013796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009261.mm.1 NM_013796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009261.mm.1 NR_027804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009261.mm.1 NR_027804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009261.mm.1 TC1600001155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009263.mm.1 NM_013796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009263.mm.1 NM_013796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009263.mm.1 NR_027804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009263.mm.1 NR_027804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009263.mm.1 TC1600001155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009265.mm.1 NR_027804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009265.mm.1 NR_027804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009265.mm.1 TC1600001155.mm.1 MTA TC
          Mouse Expression Set 430 1458344_PM_at - -
          Mouse Expression Set 430 1454718_at - -
          Mouse Expression Set 430 1454718_PM_at - -
          Mouse Expression Set 430 1458344_at - -
          Affymetrix HT Mouse Genome 430B Array 1458344_at - -
          Affymetrix HT Mouse Genome 430B Array 1458344_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454718_at - -
          Affymetrix HT Mouse Genome 430B Array 1454718_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 347842 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770053 - -
          Illumina mouse-6 v1.0 expression beadchip 1770053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770053 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2654635 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2654635 - -
          mouse-6 v1.1 (Illumina) scl48790.9.1_54-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770053 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48790.9.1_54-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2654635 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2654635 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline