Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660668_m1
          See other CTC1 GE Assays ›
          Gene Symbol:
          Ctc1
          Gene Name:
          CTS telomere maintenance complex component 1
          Gene Aliases:
          1500010J02Rik, AAF-132, AAF132
          Chromosome Location:
          Chr.11: 69015904 - 69036473 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660668_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013256.2 NP_001013274.2 11 - 12 2159 89
          NM_001143790.1 NP_001137262.1 11 - 12 2159 89
          NM_001281464.1 NP_001268393.1 11 - 12 2030 89
          NM_001281465.1 NP_001268394.1 11 - 12 2143 89
          GenBank mRNA AK038651.1 - 11 - 12 1946 89
          AK044055.1 - 11 - 12 1938 89
          AK145919.1 - 11 - 12 1974 89
          AK165906.1 - 11 - 12 1959 89
          BC014687.1 - 2 - 3 330 89
          BC060629.1 - 7 - 8 1209 6847619 89
          BC066078.1 - 11 - 12 1974 89
          BC072625.1 - 11 - 12 1987 89
          BC147495.1 - 11 - 12 1957 9056325 89
          BC147497.1 - 11 - 12 1957 9056354 89

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK038651.1, BC014687.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctc1

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          cellular response to DNA damage stimulus
          aging
          regulation of G2/M transition of mitotic cell cycle
          telomere maintenance via telomere lengthening
          multicellular organism growth
          positive regulation of DNA replication
          positive regulation of fibroblast proliferation
          spleen development
          thymus development
          bone marrow development
          chromosome organization
          hematopoietic stem cell proliferation
          replicative senescence
          DNA binding
          single-stranded DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001013256.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001013256.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001143790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001143790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001281464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 NM_001281465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007455.mm.1 TC1100000891.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline