Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00661603_m1
          See other ZFP449 GE Assays ›
          Gene Symbol:
          Zfp449
          Gene Name:
          zinc finger protein 449
          Gene Aliases:
          9530049C15Rik
          Chromosome Location:
          Chr.X: 56346383 - 56365674 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00661603_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp449
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030139.4 NP_084415.1 3 - 4 687 66
          GenBank mRNA AK020600.1 - 3 - 4 680 66
          AK036596.1 - 3 - 4 688 66
          BC082594.1 - 2 - 3 577 30642886 66
          BC145936.1 - 2 - 3 577 40131239 66
          BC145938.1 - 2 - 3 577 40131241 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000515.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp449

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32395553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32395552 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC145938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC082594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC145936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 AK036596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 AK142888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC145938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC082594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 BC145936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 AK036596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P263844 AK142888.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696802 NM_146291.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41958 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165074 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC145938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC082594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC145936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 AK036596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 AK142888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC145938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC082594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 BC145936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 AK036596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P263844 AK142888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1200510 BC082594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1200510 NM_030139.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC145938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC082594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC145936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 AK036596.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 AK142888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC145938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC082594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 BC145936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 AK036596.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263844 AK142888.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32395510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09298954 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165074 - -
          Whole Mouse Genome G4122A A_51_P263844 BC145938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 BC082594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 BC145936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 AK036596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 AK142888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 BC145938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 BC082594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 BC145936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 AK036596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263844 AK142888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1200510 BC082594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1200510 NM_030139.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122907 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165074 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09298954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15218374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15218364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15218366 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32395553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32395552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32395530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15052207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15044380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14832542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15044379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15044375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599690 - -
          Affymetrix HT MG-430 PM Array Plate 1439945_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599686 - -
          Mouse Genome 430 2.0 Array 1439945_at - -
          Mouse Genome 430 2.0 Array 1439945_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78619_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78619_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00003365.mm.1 NM_030139.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003365.mm.1 NM_030139.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003365.mm.1 TC0X00000515.mm.1 MTA TC
          Mouse Expression Set 430 1439945_at - -
          Mouse Expression Set 430 1439945_PM_at - -
          Murine Genome U74v2 Set 134597_PM_at - -
          Murine Genome U74v2 Set 134597_at - -
          Affymetrix HT Mouse Genome 430B Array 1439945_at - -
          Affymetrix HT Mouse Genome 430B Array 1439945_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 410542 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590154 - -
          Illumina mouse-6 v1.0 expression beadchip 6590154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226584 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226584 - -
          mouse-6 v1.1 (Illumina) scl54925.5_509-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54925.5_509-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226584 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline