Search
Search

Gtf2b| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145546.1 | NP_663521.1 | 2 - 3 | 151 | 151 | |
| GenBank mRNA | AK131856.1 | - | 2 - 3 | 173 | 151 | |
| AK133097.1 | - | 2 - 3 | 173 | 151 | ||
| AK133825.1 | - | 2 - 3 | 173 | 151 | ||
| AK143935.1 | - | 2 - 3 | 173 | 151 | ||
| AK145403.1 | - | 2 - 3 | 173 | 151 | ||
| AK146223.1 | - | 2 - 3 | 174 | 151 | ||
| AK146666.1 | - | 2 - 3 | 173 | 151 | ||
| AK146910.1 | - | 2 - 3 | 173 | 151 | ||
| AK151577.1 | - | 2 - 3 | 173 | 151 | ||
| BC016637.1 | - | 2 - 3 | 151 | 2650779 | 151 | |
Set Membership: |
|