Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663755_m1
          See other NPM2 GE Assays ›
          Gene Symbol:
          Npm2
          Gene Name:
          nucleophosmin/nucleoplasmin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 70647296 - 70660753 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663755_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181345.3 NP_851990.2 5 - 6 518 149
          GenBank mRNA AK054533.1 - 5 - 6 519 149
          AK139497.1 - 5 - 6 519 149
          AY262112.1 - 5 - 6 518 149
          BC048751.1 - 5 - 6 493 149
          BC104058.1 - 5 - 6 480 40040361 149
          BC104059.1 - 5 - 6 480 40040362 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002348.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npm2

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          regulation of exit from mitosis
          multicellular organism development
          single fertilization
          embryo development
          chromatin modification
          positive regulation of catalytic activity
          positive regulation of DNA replication
          positive regulation of meiotic nuclear division
          positive regulation of DNA metabolic process
          protein homooligomerization
          nucleic acid binding
          chromatin binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157220 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154526 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289679 AK054533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29702712 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154526 - -
          Whole Mouse Genome G4122A A_51_P289679 AK054533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P289679 AK054533.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135209 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11695340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11695335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11702177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11695340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11677649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11677647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11245320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11769154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11223035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11245326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11223037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11695332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11155448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11623244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11409506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11695328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29702713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10421451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421455 - -
          Affymetrix Mouse Gene 1.1 ST Array 10421444 - -
          Affymetrix Mouse Clariom S Array PSR1400018386.mm.1 NM_181345.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018386.mm.1 NM_181345.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018386.mm.1 TC1400002348.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018392.mm.1 NM_181345.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018392.mm.1 NM_181345.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018392.mm.1 TC1400002348.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 301418 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6040139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704678 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704678 - -
          mouse-6 v1.1 (Illumina) scl0328440.4_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0328440.4_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2704678 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2704678 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0