Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00723498_m1
          See other BC089491 GE Assays ›
          Gene Symbol:
          BC089491
          Gene Name:
          cDNA sequence BC089491
          Gene Aliases:
          Selv
          Chromosome Location:
          Chr.7: 28284652 - 28291134 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00723498_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC089491
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175033.3 NP_778198.2 5 - 6 1105 58
          GenBank mRNA AK077113.2 - 5 - 6 1158 58
          BC089491.1 - 5 - 6 1104 6772991 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002691.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC089491

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020990 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020922 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020988 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020532 NM_175033.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020532 AK077113.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020532 BC089491.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972767 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020532 NM_175033.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020532 AK077113.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020532 BC089491.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020984 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06149043 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140461 - -
          Whole Mouse Genome G4122A A_51_P137042 NM_175033.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137042 AK077113.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137042 BC089491.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136221 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06130057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06305554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06346630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06305548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020922 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020988 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020990 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25021035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06355656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06208847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06591060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10561373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561374 - -
          Affymetrix HT MG-430 PM Array Plate 1456100_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10561369 - -
          Mouse Genome 430 2.0 Array 1456100_at - -
          Mouse Genome 430 2.0 Array 1456100_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 280621_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 280621_at - -
          Affymetrix Mouse Clariom S Array PSR0700023472.mm.1 TC0700002691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023472.mm.1 NM_175033.4 Refseq
          Mouse Expression Set 430 1456100_PM_at - -
          Mouse Expression Set 430 1456100_at - -
          Murine Genome U74v2 Set 102746_PM_at - -
          Murine Genome U74v2 Set 102746_at - -
          Mouse Genome 430A 2.0 Array 1456100_at - -
          Mouse Genome 430A 2.0 Array 1456100_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456100_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456100_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 389468 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 510300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599891 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599891 - -
          mouse-6 v1.1 (Illumina) scl31586.7.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 510300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31586.7.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599891 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599891 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline