Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724430_m1
          See other C230052I12RIK GE Assays ›
          Gene Symbol:
          C230052I12Rik
          Gene Name:
          RIKEN cDNA C230052I12 gene
          Gene Aliases:
          AW124591, AW538696, Faap24
          Chromosome Location:
          Chr.7: 35392152 - 35396767 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724430_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C230052I12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178643.5 NP_848758.1 3 - 4 849 63
          GenBank mRNA AK031566.1 - 3 - 4 890 63
          AK040248.1 - 3 - 4 875 63
          AK082457.1 - 3 - 4 850 63
          AK153918.1 - 2 - 3 905 63
          AK166481.1 - 3 - 4 896 63
          BC096687.1 - 3 - 4 701 1433264 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002850.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C230052I12Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          cellular response to DNA damage stimulus
          biological_process
          DNA binding
          chromatin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 BC096687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK031566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK040248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK082457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 NM_178643.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 BC096687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK031566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK040248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 AK082457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P426863 NM_178643.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132560 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105722 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 BC096687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK031566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK040248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK082457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 NM_178643.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 BC096687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK031566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK040248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 AK082457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P426863 NM_178643.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25049090 - -
          Whole Mouse Genome G4122A A_52_P426863 BC096687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK031566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK040248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK082457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 NM_178643.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 BC096687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK031566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK040248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 AK082457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P426863 NM_178643.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573425 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06573427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25049096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05910309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06121312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25049088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562467 - -
          Affymetrix HT MG-430 PM Array Plate 1454779_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562461 - -
          Mouse Genome 430 2.0 Array 1454779_s_at - -
          Mouse Genome 430 2.0 Array 1454779_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101831_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101831_at - -
          Affymetrix Mouse Clariom S Array PSR0700025478.mm.1 TC0700002850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025478.mm.1 NM_178643.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025478.mm.1 NM_178643.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025482.mm.1 TC0700002850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025482.mm.1 NM_178643.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025482.mm.1 NM_178643.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025481.mm.1 TC0700002850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025481.mm.1 NM_178643.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025481.mm.1 NM_178643.5 Refseq
          Mouse Expression Set 430 1454779_PM_s_at - -
          Mouse Expression Set 430 1454779_s_at - -
          Mouse Genome 430A 2.0 Array 1454779_s_at - -
          Mouse Genome 430A 2.0 Array 1454779_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454779_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454779_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 818408 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2450576 - -
          Illumina mouse-6 v1.0 expression beadchip 2450576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216046 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216046 - -
          mouse-6 v1.1 (Illumina) scl31471.3.1_58-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31471.3.1_58-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216046 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2994631 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216046 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2994631 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline