Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724442_m1
          See other UBXN8 GE Assays ›
          Gene Symbol:
          Ubxn8
          Gene Name:
          UBX domain protein 8
          Gene Aliases:
          D0H8S2298E, Rep-8, Rep8h, Ubxd6
          Chromosome Location:
          Chr.8: 33619586 - 33643762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724442_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubxn8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178648.2 NP_848763.2 3 - 4 319 76
          GenBank mRNA AK011496.1 - 3 - 4 307 76
          AK016964.1 - 2 - 3 191 76
          AK132565.1 - 3 - 4 312 76
          AK161397.1 - 3 - 4 714 76
          BC024492.1 - 3 - 4 313 4981720 76
          D88447.1 - 3 - 4 302 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002008.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubxn8

          Gene Ontology Categories:

          Function(s) Process(es)

          ER-associated ubiquitin-dependent protein catabolic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769987 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769986 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198137 D88447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198137 BC024492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198137 D88447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198137 BC024492.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575682 AK034384.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726354 AK143276.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726354 AK145664.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726354 AY346099.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726354 BC032925.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726354 NM_178594.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63050 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137864 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101725 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198137 D88447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198137 BC024492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198137 D88447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198137 BC024492.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25769977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25770024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06472050 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137864 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06472083 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101725 - -
          Whole Mouse Genome G4122A A_51_P198137 D88447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198137 BC024492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198137 D88447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198137 BC024492.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106032 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101725 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137864 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961684 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06961690 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769985 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769987 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25769986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25769974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25770014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06961690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06961685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07042649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07135908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07042648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07109375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07396952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06939473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25769971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25770009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578113 - -
          Affymetrix HT MG-430 PM Array Plate 1454006_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571266 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578109 - -
          Mouse Genome 430 2.0 Array 1454006_a_at - -
          Mouse Genome 430 2.0 Array 1454006_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108159_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108159_at - -
          Affymetrix Mouse Clariom S Array PSR0800015749.mm.1 NM_178648.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015749.mm.1 NM_178648.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015749.mm.1 TC0800002008.mm.1 MTA TC
          Mouse Expression Set 430 1454006_PM_a_at - -
          Mouse Expression Set 430 1454006_a_at - -
          Murine Genome U74v2 Set 160803_at - -
          Murine Genome U74v2 Set 160803_PM_at - -
          Mouse Genome 430A 2.0 Array 1454006_a_at - -
          Mouse Genome 430A 2.0 Array 1454006_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454006_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454006_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 561657 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6620035 - -
          Illumina mouse-6 v1.0 expression beadchip 102320059 - -
          Illumina mouse-6 v1.0 expression beadchip 6620035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006620035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2726713 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726713 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245121 - -
          mouse-6 v1.1 (Illumina) scl000728.1_50-S - -
          mouse-6 v1.1 (Illumina) scl34948.8.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 6620035 - -
          Illumina Sentrix mouse-6.v1 bead array 102320059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34948.8.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102320059 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2726714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245121 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2726714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline