Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00725194_m1
          See other GPR142 GE Assays ›
          Gene Symbol:
          Gpr142
          Gene Name:
          G protein-coupled receptor 142
          Gene Aliases:
          PGR2
          Chromosome Location:
          Chr.11: 114796965 - 114806808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00725194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr142
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181749.1 NP_861414.1 1 - 2 94 62
          GenBank mRNA AY288428.1 - 1 - 2 94 62
          AY957501.1 - 1 - 2 94 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001879.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr142

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051239 AY957501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051239 NM_181749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051239 AY288428.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459893 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130110 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051239 AY957501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051239 NM_181749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051239 AY288428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P248804 AY957501.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P248804 NM_181749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P248804 AY288428.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28149851 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07500805 - -
          Whole Mouse Genome G4122A A_52_P248804 AY957501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P248804 NM_181749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P248804 AY288428.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112603 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28149856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09024421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09027582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09627702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09664226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09422138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09111077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09147202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09390183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09422135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09230259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09111076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09736022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28149925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382422 - -
          Affymetrix Mouse Gene 1.1 ST Array 10382421 - -
          Affymetrix Mouse Clariom S Array PSR1100017600.mm.1 NM_181749.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017600.mm.1 NM_181749.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017600.mm.1 TC1100001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017601.mm.1 NM_181749.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017601.mm.1 NM_181749.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017601.mm.1 TC1100001879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017603.mm.1 NM_181749.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017603.mm.1 NM_181749.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017603.mm.1 TC1100001879.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 807444 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190148 - -
          Illumina mouse-6 v1.0 expression beadchip 2190148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214518 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214518 - -
          mouse-6 v1.1 (Illumina) scl40743.3.1_135-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40743.3.1_135-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214518 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214518 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline