Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00726089_s1
          See other SRM GE Assays ›
          Gene Symbol:
          Srm
          Gene Name:
          spermidine synthase
          Gene Aliases:
          AA407669, SpdST, SpdSy
          Chromosome Location:
          Chr.4: 148591513 - 148594619 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00726089_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009272.4 NP_033298.1 7 1278 74
          GenBank mRNA BC005566.1 - 7 1282 3966710 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srm

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          polyamine metabolic process
          polyamine biosynthetic process
          spermidine biosynthetic process
          catalytic activity
          spermidine synthase activity
          transferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395333 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395335 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395332 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 L19311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK051101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 NM_009272.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK028359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 L19311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK051101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 NM_009272.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK028359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002376 AK051101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002376 NM_009272.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002376 AK028359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002376 BC005566.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 AK005582.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 AK018833.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 AK018851.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 BC049608.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 BC059728.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 M20625.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 NM_009989.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906900 X55771.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151418 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002376 AK051101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002376 NM_009272.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002376 AK028359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002376 BC005566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 L19311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK051101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 NM_009272.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK028359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK146395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 L19311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK051101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 NM_009272.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK028359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 L19311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK051101.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 NM_009272.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK028359.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 L19311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK051101.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 NM_009272.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK028359.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23395344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151418 - -
          Whole Mouse Genome G4122A A_51_P300709 L19311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK051101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 NM_009272.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK028359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK146395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 L19311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK051101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 NM_009272.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK028359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300709 AK146395.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03643843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03643842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03643844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03453849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395335 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395333 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23395332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23395404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03789036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03426113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03643842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03453850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03453857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23395325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03514181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03453858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510397 - -
          Affymetrix HT MG-430 PM Array Plate 1421260_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510391 - -
          Mouse Genome 430 2.0 Array 1421260_a_at - -
          Mouse Genome 430 2.0 Array 1421260_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20810_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20810_at - -
          Affymetrix Murine Genome 6500A Array Msa13230_at - -
          Mouse Expression Set 430 1421260_PM_a_at - -
          Mouse Expression Set 430 1421260_a_at - -
          Murine Genome U74v2 Set 92540_PM_f_at - -
          Murine Genome U74v2 Set 92540_f_at - -
          Murine Genome U74v2 Set 168844_at - -
          Murine Genome U74v2 Set 168844_PM_at - -
          Mouse Genome 430A 2.0 Array 1421260_a_at - -
          Mouse Genome 430A 2.0 Array 1421260_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421260_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421260_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 901980 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850097 - -
          Illumina mouse-6 v1.0 expression beadchip 1850097 - -
          Illumina mouse-6 v1.0 expression beadchip 2480162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2630641 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225880 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225880 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2630641 - -
          mouse-6 v1.1 (Illumina) scl020810.7_29-S - -
          mouse-6 v1.1 (Illumina) scl0002839.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480162 - -
          Illumina Sentrix mouse-6.v1 bead array 1850097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002839.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020810.7_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850097 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2630641 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225880 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2809611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2809611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225880 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2630641 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002839.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020810.7_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0