Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00726901_s1
          See other DAZAP2 GE Assays ›
          Gene Symbol:
          Dazap2
          Gene Name:
          DAZ associated protein 2
          Gene Aliases:
          AI314727, Brbp, Gcap28, Prtb, gt6-12, mKIAA0058
          Chromosome Location:
          Chr.15: 100615662 - 100620761 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00726901_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dazap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011873.2 NP_036003.2 3 254 56
          GenBank mRNA AF085348.1 - 3 219 56
          AK002595.1 - 3 291 56
          AK050159.1 - 3 268 56
          AK088916.1 - 3 276 56
          AK132139.1 - 3 268 56
          AK137246.1 - 3 255 56
          AK145398.1 - 3 288 56
          AK146970.1 - 3 279 56
          AK151730.1 - 1 1862 56
          AK152511.1 - 1 1862 56
          AK169769.1 - 1 1444 56
          BC014808.1 - 3 254 4506124 56
          BC029351.1 - 2 185 56
          BC083347.1 - 3 246 6829350 56

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dazap2

          Gene Ontology Categories:

          Process(es)

          protein binding
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486709 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486712 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486714 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486710 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486706 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486713 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486703 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486705 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136378 BC014808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136378 BC029351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136378 BC083347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136378 AK145398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136378 AK146970.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61045 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100993 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136378 BC014808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136378 BC029351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136378 BC083347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136378 AK145398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136378 AK146970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321580 AF085348.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30486709 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08482004 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100993 - -
          Whole Mouse Genome G4122A A_51_P321580 AF085348.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139875 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12460495 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12304269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12460492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12460497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486713 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486710 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486712 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486705 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486709 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486714 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486706 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30486703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30486781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12460492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12326606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12037999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11895557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12460497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11872545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12183610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30486718 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10426923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426922 - -
          Affymetrix HT MG-430 PM Array Plate 1415796_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10426921 - -
          Mouse Genome 430 2.0 Array 1415796_at - -
          Mouse Genome 430 2.0 Array 1415796_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23994_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23994_at - -
          Mouse Expression Set 430 1415796_PM_at - -
          Mouse Expression Set 430 1415796_at - -
          Murine Genome U74v2 Set 94454_at - -
          Murine Genome U74v2 Set 94454_PM_at - -
          Murine Genome U74v2 Set 138406_PM_f_at - -
          Murine Genome U74v2 Set 138406_f_at - -
          Mouse Genome 430A 2.0 Array 1415796_PM_at - -
          Mouse Genome 430A 2.0 Array 1415796_at - -
          Affymetrix HT Mouse Genome 430A Array 1415796_at - -
          Affymetrix HT Mouse Genome 430A Array 1415796_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753658 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6200102 - -
          Illumina mouse-6 v1.0 expression beadchip 6200102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739287 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2739287 - -
          mouse-6 v1.1 (Illumina) scl023994.4_204-S - -
          Illumina Sentrix mouse-6.v1 bead array 6200102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl023994.4_204-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739287 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739287 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl023994.4_204-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline